Product Details
- SNP ID
-
rs401442
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:29765536 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCCTCTCTTTTCTCCCGCTGCAC[C/T]TGCCAGCCCGTCTGACAGCACTGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZG16
PubMed Links
Gene Details
- Gene
- ZG16
- Gene Name
- zymogen granule protein 16
View Full Product Details