Product Details

SNP ID
rs565105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:172444327 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATTTCATATAAGGTGAATGATAA[G/T]TTTTGTATATACTCTAGAGCTGTAC
Phenotype
MIM: 601898
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GHSR PubMed Links

Gene Details

Gene
GHSR
Gene Name
growth hormone secretagogue receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004122.2 1978 Intron NP_004113.1
NM_198407.2 1978 UTR 3 NP_940799.1

View Full Product Details