Product Details

SNP ID
rs196448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8806077 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATTGTTGATACTGGAATGTCACT[C/T]GTGACCCTCCAAAAATACCTGGAGA
Phenotype
MIM: 611462
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MFSD6L PubMed Links

Gene Details

Gene
MFSD6L
Gene Name
major facilitator superfamily domain containing 6 like
There are no transcripts associated with this gene.

Gene
PIK3R6
Gene Name
phosphoinositide-3-kinase regulatory subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010855.3 Intron NP_001010855.1
NM_001290211.1 Intron NP_001277140.1
XM_011523677.2 Intron XP_011521979.1
XM_011523678.2 Intron XP_011521980.1
XM_011523680.2 Intron XP_011521982.1
XM_011523681.2 Intron XP_011521983.1
XM_011523683.2 Intron XP_011521985.1
XM_011523686.2 Intron XP_011521988.1
XM_017024228.1 Intron XP_016879717.1
XM_017024229.1 Intron XP_016879718.1
XM_017024230.1 Intron XP_016879719.1
XM_017024231.1 Intron XP_016879720.1

View Full Product Details