Product Details

SNP ID
rs108885
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:14217619 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATATTATTGATTTTAAGACTTT[C/T]AGGTTCATCCTTTTAATTCATCTTT
Phenotype
MIM: 609252
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LIPI PubMed Links

Gene Details

Gene
LIPI
Gene Name
lipase I
There are no transcripts associated with this gene.

Gene
RBM11
Gene Name
RNA binding motif protein 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320602.1 Intron NP_001307531.1
NM_144770.4 Intron NP_658983.3
XM_005260997.4 Intron XP_005261054.1
XM_017028384.1 Intron XP_016883873.1
XM_017028385.1 Intron XP_016883874.1
XM_017028386.1 Intron XP_016883875.1
XM_017028387.1 Intron XP_016883876.1

View Full Product Details