Product Details

SNP ID
rs2032611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:19704538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTATAGTTCCCTATGACAGTATC[G/A]AAGAAAGAAAGAGGTGAAAAGAGTA
Phenotype
MIM: 426000
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
KDM5D PubMed Links

Gene Details

Gene
KDM5D
Gene Name
lysine demethylase 5D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146705.1 6542 Intron NP_001140177.1
NM_001146706.1 6542 Intron NP_001140178.1
NM_004653.4 6542 Intron NP_004644.2
XM_005262560.2 6542 UTR 3 XP_005262617.1
XM_005262561.2 6542 UTR 3 XP_005262618.1
XM_011531468.2 6542 UTR 3 XP_011529770.1

View Full Product Details