Product Details
- SNP ID
-
rs268684
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:40425001 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCTGCTTCGAGGAAGACTGTGGGG[C/G]GCAAAAAGAGGGGGGATCTGAGACC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SERTAD1
PubMed Links
Gene Details
- Gene
- SERTAD1
- Gene Name
- SERTA domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_013376.3 |
|
Intron |
|
|
NP_037508.2 |
View Full Product Details