Product Details

SNP ID
rs558718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7844997 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGGCTGTGGGAAGGTGCCCAGTG[A/G]CCCAACGGTAGTCAGGCAGGTATAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EVI5L PubMed Links

Gene Details

Gene
EVI5L
Gene Name
ecotropic viral integration site 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159944.2 Intron NP_001153416.1
NM_145245.4 Intron NP_660288.1
XM_005272458.4 Intron XP_005272515.1
XM_017026239.1 Intron XP_016881728.1

View Full Product Details