Product Details

SNP ID
rs983725326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34366816 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGGCGTGGAGGCCGCCCGGGAG[C/T]GGATGTTCAATGGTGAGAAGATCAA
Phenotype
MIM: 172400
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPI PubMed Links

Gene Details

Gene
GPI
Gene Name
glucose-6-phosphate isomerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184722.1 Intron NP_001171651.1
NM_001289789.1 Intron NP_001276718.1

View Full Product Details