Product Details

SNP ID
rs35151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:64944782 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGAATTGCCACAGCTGGCCTAAT[C/T]TCCGCTCACACTCTATATCTCACCA
Phenotype
MIM: 600023
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDH11 PubMed Links

Gene Details

Gene
CDH11
Gene Name
cadherin 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308392.1 5767 UTR 3 NP_001295321.1
NM_001797.3 5767 UTR 3 NP_001788.2
XM_005255761.3 5767 Intron XP_005255818.1
XM_005255762.2 5767 Intron XP_005255819.1
XM_005255763.2 5767 Intron XP_005255820.1
XM_011522803.2 5767 Intron XP_011521105.1
XM_017022830.1 5767 Intron XP_016878319.1

View Full Product Details