Product Details
- SNP ID
-
rs11813643
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:92419485 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- ACTGTGAGGGGTCCATGGATGAATG[A/C]GAGTCATGAAGGTGAGGAACTTAAG
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MARK2P9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144392077] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MARK2P9
- Gene Name
- microtubule affinity regulating kinase 2 pseudogene 9
There are no transcripts associated with this gene.
View Full Product Details