Product Details
- SNP ID
-
rs873711
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:2846822 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GCCCTGCCACAGAAATTTCTTCACT[A/G]TTTCCAGCAGGGGACTGCAATTGAG
- Phenotype
-
MIM: 608550
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PCED1A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114556558] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PCED1A
- Gene Name
- PC-esterase domain containing 1A
There are no transcripts associated with this gene.
- Gene
- VPS16
- Gene Name
- VPS16, CORVET/HOPS core subunit
View Full Product Details