Product Details

SNP ID
rs2256518
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:37009164 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGTTGCTTTATACTTACTGATGTA[A/C]TGACTTGCTTTCATAGAATTTACCT
Phenotype
MIM: 609892
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RIPPLY3 PubMed Links

Gene Details

Gene
RIPPLY3
Gene Name
ripply transcriptional repressor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317768.1 Intron NP_001304697.1
NM_001317777.1 Intron NP_001304706.1
NM_018962.2 Intron NP_061835.1

View Full Product Details