Product Details
- SNP ID
-
rs1059004
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:33028155 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTAGAACTCATCCGACCCCCGACC[A/C]CCACCTCCGGGAAAAGATTCTAAAA
- Phenotype
-
MIM: 606386
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
OLIG2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6517136] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OLIG2
- Gene Name
- oligodendrocyte lineage transcription factor 2
View Full Product Details