Product Details
- SNP ID
-
rs2519459
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:21104515 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGAAGAAACTCTCCTGTCCCCATTG[G/T]GTCTTTCTGATCTCTGGTGCCGATT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
BCRP2
PubMed Links
Gene Details
- Gene
- BCRP2
- Gene Name
- breakpoint cluster region pseudogene 2
There are no transcripts associated with this gene.
View Full Product Details