Product Details

SNP ID
rs9627731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:49418499 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTCTGGCCGGGCAGGCTGGAACC[C/T]GGGCTTGTTGTGGGCCGCAGACCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C22orf34 PubMed Links

Gene Details

Gene
C22orf34
Gene Name
chromosome 22 open reading frame 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289922.2 Intron NP_001276851.1
XM_011530167.2 Intron XP_011528469.1
XM_017028796.1 Intron XP_016884285.1

View Full Product Details