Product Details

SNP ID
rs8180638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:16134945 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATATTCAAGGAAGGTTTTTAAAA[C/T]GACATGTAAAATTAAATCTGAGCAT
Phenotype
MIM: 610082
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4639 PubMed Links

Gene Details

Gene
MIR4639
Gene Name
microRNA 4639
There are no transcripts associated with this gene.

Gene
MYLIP
Gene Name
myosin regulatory light chain interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013262.3 Intron NP_037394.2
XM_005249032.2 Intron XP_005249089.1
XM_005249033.2 Intron XP_005249090.1
XM_017010789.1 Intron XP_016866278.1

View Full Product Details