Product Details
- SNP ID
-
rs4820192
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:35381991 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGAAAGACTTTTTCTTTGACTAA[A/C]GCAGACTTTCTCAGGCTTTTTAACC
- Phenotype
-
MIM: 141250
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HMOX1
PubMed Links
Gene Details
- Gene
- HMOX1
- Gene Name
- heme oxygenase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002133.2 |
|
Intron |
|
|
NP_002124.1 |
View Full Product Details