Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001270393.1 | Intron | NP_001257322.1 | |||
NM_001270394.1 | Intron | NP_001257323.1 | |||
NM_001270395.1 | Intron | NP_001257324.1 | |||
NM_032471.5 | Intron | NP_115860.1 | |||
NM_181794.2 | Intron | NP_861459.1 | |||
NM_181795.2 | Intron | NP_861460.1 | |||
XM_011535930.2 | Intron | XP_011534232.1 | |||
XM_011535931.2 | Intron | XP_011534233.1 | |||
XM_011535932.2 | Intron | XP_011534234.1 | |||
XM_011535933.2 | Intron | XP_011534235.1 | |||
XM_011535934.2 | Intron | XP_011534236.1 | |||
XM_011535935.2 | Intron | XP_011534237.1 | |||
XM_011535936.1 | Intron | XP_011534238.1 | |||
XM_011535937.2 | Intron | XP_011534239.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_020755.3 | Intron | NP_065806.1 |