Product Details
- SNP ID
-
rs2413426
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:36861813 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTTGATCGCTGCTGTATCACTAG[A/G]AGGTAGCTCATCCCTATTGAGCACC
- Phenotype
-
MIM: 601488
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NCF4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11912791] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NCF4
- Gene Name
- neutrophil cytosolic factor 4
View Full Product Details