Product Details

SNP ID
rs1233664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28217948 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTGGGATTGAGAGGCCGGTTGGC[C/T]GGGATGAAGTTCAATGAATGCACAG
Phenotype
MIM: 602246
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOB2P1 PubMed Links

Gene Details

Gene
TOB2P1
Gene Name
transducer of ERBB2, 2 pseudogene 1
There are no transcripts associated with this gene.

Gene
ZSCAN9
Gene Name
zinc finger and SCAN domain containing 9
There are no transcripts associated with this gene.

View Full Product Details