Product Details

SNP ID
rs1076608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:45985679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCCTTCACGAGGCCTCAGGATC[C/G]CAGTGGGGGTACCCGAGGCTGCCCT
Phenotype
MIM: 120220
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COL6A1 PubMed Links

Gene Details

Gene
COL6A1
Gene Name
collagen type VI alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001848.2 Intron NP_001839.2

View Full Product Details