Product Details

SNP ID
rs705581
Assay Type
Validated
NCBI dbSNP Submissions
50
Location
Chr.1:15114998 on Build GRCh38
Set Membership
JSNP Validated
Context Sequence [VIC/FAM]
CTTAAACGGAGCTGCCAAGGTGGGA[A/G]GAGGCCCACAGTTCCCTAAAACACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KAZN PubMed Links

Gene Details

Gene
KAZN
Gene Name
kazrin, periplakin interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017999.2 4964 Intron NP_001017999.1
NM_001018000.3 4964 Intron NP_001018000.1
NM_001018001.2 4964 Intron NP_001018001.1
NM_015209.2 4964 Intron NP_056024.1
NM_201628.2 4964 UTR 3 NP_963922.2
XM_005245795.4 4964 UTR 3 XP_005245852.1
XM_011541074.2 4964 UTR 3 XP_011539376.1
XM_011541075.2 4964 UTR 3 XP_011539377.1
XM_011541076.2 4964 UTR 3 XP_011539378.1
XM_011541077.2 4964 UTR 3 XP_011539379.1
XM_011541080.2 4964 Intron XP_011539382.1
XM_011541081.2 4964 UTR 3 XP_011539383.1
XM_017000768.1 4964 Intron XP_016856257.1
XM_017000769.1 4964 Intron XP_016856258.1
XM_017000770.1 4964 Intron XP_016856259.1
XM_017000771.1 4964 Intron XP_016856260.1
XM_017000772.1 4964 Intron XP_016856261.1
Gene
TMEM51-AS1
Gene Name
TMEM51 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details