Product Details

SNP ID
rs557911
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:107870178 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATGAAGTAAATGAAAAAAATCCTC[A/G]GTGACACTGCCTCCTGCCTGCCAAG
Phenotype
MIM: 608648
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SEC63 PubMed Links

Gene Details

Gene
SEC63
Gene Name
SEC63 homolog, protein translocation regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007214.4 4078 UTR 3 NP_009145.1
XM_011535399.2 4078 Intron XP_011533701.1
XM_017010218.1 4078 Intron XP_016865707.1

View Full Product Details