Product Details

SNP ID
rs6933329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:107868207 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATGGAGTTAAATCAGACCGTTTG[C/T]GGGAAAATTGTTTCTTAAAAAAAAA
Phenotype
MIM: 608648
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEC63 PubMed Links

Gene Details

Gene
SEC63
Gene Name
SEC63 homolog, protein translocation regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007214.4 6049 UTR 3 NP_009145.1
XM_011535399.2 6049 Intron XP_011533701.1
XM_017010218.1 6049 Intron XP_016865707.1

View Full Product Details