Product Details

SNP ID
rs4820295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37687094 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCATCAGGAAACTCCTAGCGTGCA[C/T]CAACAGGCCCTACAGGCGAGTTTAG
Phenotype
MIM: 150570 MIM: 609761
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LGALS1 PubMed Links

Gene Details

Gene
LGALS1
Gene Name
galectin 1
There are no transcripts associated with this gene.

Gene
NOL12
Gene Name
nucleolar protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024313.2 Intron NP_077289.1
Gene
TRIOBP
Gene Name
TRIO and F-actin binding protein
There are no transcripts associated with this gene.

View Full Product Details