Product Details

SNP ID
rs12599176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1447136 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCCGCCCACACAGCAGAGGCAGG[C/T]GGGACCCTCACCCAAGCTGGCTCCC
Phenotype
MIM: 602727
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC154 PubMed Links

Gene Details

Gene
CCDC154
Gene Name
coiled-coil domain containing 154
There are no transcripts associated with this gene.

Gene
CLCN7
Gene Name
chloride voltage-gated channel 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114331.2 Intron NP_001107803.1
NM_001287.5 Intron NP_001278.1
XM_011522354.1 Intron XP_011520656.1

View Full Product Details