Product Details

SNP ID
rs11583790
Assay Type
Validated
NCBI dbSNP Submissions
30
Location
Chr.1:22063585 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TAGAGAGGGTCTGTTAGGTGCTTGG[A/G]ATCTTGTTCTTCAACTGCACACATA
Phenotype
MIM: 116952
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CDC42 PubMed Links
Additional Information
For this assay, SNP(s) [rs10158376] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CDC42
Gene Name
cell division cycle 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039802.1 Intron NP_001034891.1
NM_001791.3 Intron NP_001782.1
NM_044472.2 Intron NP_426359.1

View Full Product Details