Product Details

SNP ID
rs7740351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28431635 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCATTGGCAGTTGGCATAGAAAA[A/C]TGGGGATAGGTTCAAGGAGATCTAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZSCAN23 PubMed Links

Gene Details

Gene
ZSCAN23
Gene Name
zinc finger and SCAN domain containing 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012455.1 1779 Intron NP_001012458.1
XM_005248950.2 1779 UTR 3 XP_005249007.1
XM_011514406.2 1779 Intron XP_011512708.1
XM_011514408.2 1779 Intron XP_011512710.1
XM_011514410.2 1779 Intron XP_011512712.1
XM_017010479.1 1779 Intron XP_016865968.1

View Full Product Details