Product Details

SNP ID
rs1294028
Assay Type
Validated
NCBI dbSNP Submissions
77
Location
Chr.1:9304575 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGCATCTCCAAGGTGGCATGGGTA[A/G]GCTTCGGTAACTAGTTGAATGTGGG
Phenotype
MIM: 611657
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPSB1 PubMed Links

Gene Details

Gene
SPSB1
Gene Name
splA/ryanodine receptor domain and SOCS box containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025106.3 Intron NP_079382.2

View Full Product Details