Product Details

SNP ID
rs736793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41030180 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGATTCACTGTTAAATGTTAATTG[A/T]GCACCTACTATGCACCAGGCATCAT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929555 PubMed Links

Gene Details

Gene
LOC101929555
Gene Name
uncharacterized LOC101929555
There are no transcripts associated with this gene.

Gene
UNC5CL
Gene Name
unc-5 family C-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173561.2 Intron NP_775832.2

View Full Product Details