Product Details

SNP ID
rs71325268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:27929647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTGGACTGGACTGGCATACTTA[C/T]CTTGATCTTGACTTTAAGATGCTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR3199-1 PubMed Links

Gene Details

Gene
MIR3199-1
Gene Name
microRNA 3199-1
There are no transcripts associated with this gene.

Gene
MIR3199-2
Gene Name
microRNA 3199-2
There are no transcripts associated with this gene.

Gene
TTC28-AS1
Gene Name
TTC28 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details