Product Details

SNP ID
rs10779506
Assay Type
Functionally tested
NCBI dbSNP Submissions
51
Location
Chr.1:209430015 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACCTGTACTGGAAAATTTAGGAC[C/G]TGGTTTCATTCTTTGAGTCTCATGT
Phenotype
MIM: 613147
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR205 PubMed Links

Gene Details

Gene
MIR205
Gene Name
microRNA 205
There are no transcripts associated with this gene.

Gene
MIR205HG
Gene Name
MIR205 host gene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104548.1 Intron NP_001098018.1
XM_017002058.1 Intron XP_016857547.1

View Full Product Details