Product Details

SNP ID
rs2249632
Assay Type
Validated
NCBI dbSNP Submissions
57
Location
Chr.1:209431757 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
GTTTCCTCTAGAAAGGACCAACCAC[A/G]CCGAGCTCAGTTATGGCACACACAG
Phenotype
MIM: 613147
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MIR205 PubMed Links
Additional Information
For this assay, SNP(s) [rs79238475] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MIR205
Gene Name
microRNA 205
There are no transcripts associated with this gene.

Gene
MIR205HG
Gene Name
MIR205 host gene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104548.1 Intron NP_001098018.1
XM_017002058.1 Intron XP_016857547.1

View Full Product Details