Product Details

SNP ID
rs6138435
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:24964945 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AATTCCCAAGGCTTCCCACTACCTA[C/G]GAAATGCCATCTCCTGCTGTTCGGC
Phenotype
MIM: 615884 MIM: 603253
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APMAP PubMed Links

Gene Details

Gene
APMAP
Gene Name
adipocyte plasma membrane associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020531.2 Intron NP_065392.1
XM_005260763.3 Intron XP_005260820.1
Gene
CST7
Gene Name
cystatin F
There are no transcripts associated with this gene.

View Full Product Details