Product Details

SNP ID
rs3761996
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:143059965 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTCCATTAAGCCCTGGTAGTGTAG[C/T]GTAAGAAAAAAGTAAGGGCAAAACC
Phenotype
MIM: 608514
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIG1 PubMed Links

Gene Details

Gene
AIG1
Gene Name
androgen induced 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286587.1 Intron NP_001273516.1
NM_001286588.1 Intron NP_001273517.1
NM_001286589.1 Intron NP_001273518.1
NM_016108.3 Intron NP_057192.2
XM_011535881.2 Intron XP_011534183.1
XM_011535882.2 Intron XP_011534184.1
XM_011535883.2 Intron XP_011534185.1
XM_011535884.2 Intron XP_011534186.1
XM_011535885.2 Intron XP_011534187.1
XM_017010927.1 Intron XP_016866416.1
XM_017010928.1 Intron XP_016866417.1

View Full Product Details