Product Details

SNP ID
rs228729
Assay Type
Validated
NCBI dbSNP Submissions
92
Location
Chr.1:7785635 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CATCCTACGAATGCACCAGGACTCA[C/T]ACAAGCAGCCAGAGGAGTGGTCAGT
Phenotype
MIM: 603427 MIM: 603657
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PER3 PubMed Links

Gene Details

Gene
PER3
Gene Name
period circadian clock 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289861.1 Intron NP_001276790.1
NM_001289862.1 Intron NP_001276791.1
NM_001289863.1 Intron NP_001276792.1
NM_001289864.1 Intron NP_001276793.1
NM_016831.2 Intron NP_058515.1
XM_005263524.3 Intron XP_005263581.1
XM_011542385.2 Intron XP_011540687.1
XM_011542390.2 Intron XP_011540692.1
XM_017002723.1 Intron XP_016858212.1
XM_017002724.1 Intron XP_016858213.1
XM_017002725.1 Intron XP_016858214.1
XM_017002726.1 Intron XP_016858215.1
XM_017002727.1 Intron XP_016858216.1
XM_017002728.1 Intron XP_016858217.1
XM_017002729.1 Intron XP_016858218.1
XM_017002730.1 Intron XP_016858219.1
XM_017002731.1 Intron XP_016858220.1
XM_017002732.1 Intron XP_016858221.1
XM_017002733.1 Intron XP_016858222.1
XM_017002734.1 Intron XP_016858223.1
XM_017002735.1 Intron XP_016858224.1
XM_017002736.1 Intron XP_016858225.1
XM_017002737.1 Intron XP_016858226.1
XM_017002738.1 Intron XP_016858227.1
Gene
VAMP3
Gene Name
vesicle associated membrane protein 3
There are no transcripts associated with this gene.

View Full Product Details