Product Details
- SNP ID
-
rs12195744
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:163339550 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTTTTGAGATATAAGAATAATAGA[A/G]AGTGAATTAGCGGATTAAAAGATCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DKFZp451B082
PubMed Links
Gene Details
- Gene
- DKFZp451B082
- Gene Name
- uncharacterized LOC401282
There are no transcripts associated with this gene.
View Full Product Details