Product Details

SNP ID
rs926587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:4610860 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTGTTGGGAGGCAAGTGTGTGAC[A/T]GGTGTGCAAGTCAGGTCAGGAGGCT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KU-MEL-3 PubMed Links

Gene Details

Gene
KU-MEL-3
Gene Name
uncharacterized LOC497048
There are no transcripts associated with this gene.

Gene
LOC105374896
Gene Name
uncharacterized LOC105374896
There are no transcripts associated with this gene.

View Full Product Details