Product Details

SNP ID
rs6675059
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:40250629 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCTCTGCCACTTCCAATATATAA[C/G]CTTGGACAATGACAGAGTTCAAACT
Phenotype
MIM: 180610 MIM: 606480
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RLF PubMed Links

Gene Details

Gene
RLF
Gene Name
rearranged L-myc fusion
There are no transcripts associated with this gene.

Gene
TMCO2
Gene Name
transmembrane and coiled-coil domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008740.3 Intron NP_001008740.1
Gene
ZMPSTE24
Gene Name
zinc metallopeptidase STE24
There are no transcripts associated with this gene.

View Full Product Details