Product Details

SNP ID
rs9614014
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:29622211 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GCATTTGGTGTATGATACTATCTCA[A/G]AAGCATTTTGTAAACTTGCCAGTGG
Phenotype
MIM: 607379
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NF2 PubMed Links

Gene Details

Gene
NF2
Gene Name
neurofibromin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000268.3 Intron NP_000259.1
NM_016418.5 Intron NP_057502.2
NM_181825.2 Intron NP_861546.1
NM_181828.2 Intron NP_861966.1
NM_181829.2 Intron NP_861967.1
NM_181830.2 Intron NP_861968.1
NM_181831.2 Intron NP_861969.1
NM_181832.2 Intron NP_861970.1
NM_181833.2 Intron NP_861971.1
XM_017028809.1 Intron XP_016884298.1
XM_017028810.1 Intron XP_016884299.1

View Full Product Details