Product Details

SNP ID
rs3820185
Assay Type
Functionally tested
NCBI dbSNP Submissions
70
Location
Chr.1:186839211 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TGTGCGGGTCAAGTCAGGCAGCATG[G/T]TGAGAAGAGAGAGTGTTTTTTGTGG
Phenotype
MIM: 600522
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PLA2G4A PubMed Links

Gene Details

Gene
PLA2G4A
Gene Name
phospholipase A2 group IVA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001311193.1 Intron NP_001298122.1
NM_024420.2 Intron NP_077734.1
XM_005245267.3 Intron XP_005245324.2
XM_011509642.2 Intron XP_011507944.1

View Full Product Details