Product Details
- SNP ID
-
rs232280
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45712889 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCAGAAAAGTTCAATCAAATCTTCA[A/G]GGAATAAACCATTTCAATCTTATAT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR3617
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76519666] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR3617
- Gene Name
- microRNA 3617
There are no transcripts associated with this gene.
- Gene
- SPINT4
- Gene Name
- serine peptidase inhibitor, Kunitz type 4
There are no transcripts associated with this gene.
- Gene
- WFDC10B
- Gene Name
- WAP four-disulfide core domain 10B
There are no transcripts associated with this gene.
- Gene
- WFDC13
- Gene Name
- WAP four-disulfide core domain 13
There are no transcripts associated with this gene.
- Gene
- WFDC3
- Gene Name
- WAP four-disulfide core domain 3
View Full Product Details