Product Details

SNP ID
rs6100209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:36542744 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACTCCACTCCCCTTCTGGGGCCT[C/G]TTTTCCTAGCTGCACCATGGGGGCA
Phenotype
MIM: 609905
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DLGAP4-AS1 PubMed Links

Gene Details

Gene
DLGAP4-AS1
Gene Name
DLGAP4 antisense RNA 1
There are no transcripts associated with this gene.

Gene
MYL9
Gene Name
myosin light chain 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006097.4 Intron NP_006088.2
NM_181526.2 Intron NP_852667.1

View Full Product Details