Product Details
- SNP ID
-
rs12559811
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:37604977 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAATGAGAGATGCATTGAGATGTGT[G/T]GATGTGTGTTCTTGGTTTCTACATT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LANCL3
PubMed Links
Gene Details
- Gene
- LANCL3
- Gene Name
- LanC like 3
View Full Product Details