Product Details

SNP ID
rs6930
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:95583747 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACATTTCTTTTTTCTAGTGTTTTA[A/G]AGATAGTTCACAGTATTTGAGTTAA
Phenotype
MIM: 602527
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PDK4 PubMed Links

Gene Details

Gene
PDK4
Gene Name
pyruvate dehydrogenase kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002612.3 3450 UTR 3 NP_002603.1

View Full Product Details