Product Details

SNP ID
rs12369492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110470449 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCACAGTCCTTGGCCGACTGCAGG[A/C]TCCTGGTGTATCAGGAGATACGTGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM216A PubMed Links

Gene Details

Gene
FAM216A
Gene Name
family with sequence similarity 216 member A
There are no transcripts associated with this gene.

Gene
GPN3
Gene Name
GPN-loop GTPase 3
There are no transcripts associated with this gene.

View Full Product Details