Product Details
- SNP ID
-
rs4806174
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:35534044 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTTGGAACCTTGCCGGGGACGTGCT[C/G]ACCACTTCGCAGGCGAGAGCTCAGG
- Phenotype
-
MIM: 609169
MIM: 609969
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GAPDHS
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115953720] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GAPDHS
- Gene Name
- glyceraldehyde-3-phosphate dehydrogenase, spermatogenic
- Gene
- SBSN
- Gene Name
- suprabasin
There are no transcripts associated with this gene.
- Gene
- TMEM147-AS1
- Gene Name
- TMEM147 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details