Product Details

SNP ID
rs12532771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:17302172 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGCTTACCTGTACATAGAAAGTT[A/G]GCACTGAATGTTGGCAAATAATAGG
Phenotype
MIM: 600253
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AHR PubMed Links

Gene Details

Gene
AHR
Gene Name
aryl hydrocarbon receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001621.4 Intron NP_001612.1

View Full Product Details