Product Details
- SNP ID
-
rs35664355
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:2380385 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAGCCTGACCTCTTTCCCAGGAAG[A/G]AGCCCACACACACACGCACAGGCAC
- Phenotype
-
MIM: 186845
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CD81
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116794482] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CD81
- Gene Name
- CD81 molecule
- Gene
- CD81-AS1
- Gene Name
- CD81 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details