Product Details
- SNP ID
-
rs7221765
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40876731 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTCAAATTTTGTCTTGTTTTTCCT[C/T]TTCAGGATGTCAGGATGTTTCAAGA
- Phenotype
-
MIM: 601687
MIM: 608218
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KRT12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114253486] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRT12
- Gene Name
- keratin 12
There are no transcripts associated with this gene.
- Gene
- KRT20
- Gene Name
- keratin 20
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019010.2 |
|
Intron |
|
|
NP_061883.1 |
View Full Product Details